missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies BrainPhys™ Imaging Optimized Medium

Product Code. 17138301
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17138301 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17138301 Supplier STEMCELL Technologies Supplier No. 05796
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Serum-free neurophysiological basal medium for improved neuronal live imaging and function

  • BrainPhys™ Imaging Optimized Medium is a serum-free neuronal basal medium (based on the BrainPhys™ formulation published by Cedric Bardy and Fred H.Gage) that is optimized for imaging applications
  • Applications of BrainPhys™ Imaging Optimized Medium include live fluorescent imaging (calcium imaging and optogenetics) and neuronal culture
  • In addition to the removal of phenol red, the formulation has been modified to reduce background fluorescence and increase stability upon repeated exposure to light (M Zabolocki et al.) Nat Comm, 2020)

Specifications

Cell Type Neural Cells, PSC-Derived, Neurons, Pluripotent Stem Cells
For Use With (Application) Cell Culture; Functional Assay; Immunofluorescence; Organoid Culture
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.