missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Four-Clear Sided Disposable Cuvettes for Fluorescence

Product Code. 15594789 Shop All Brand Products
Change view
Click to view available options
Quantity:
100
500
Unit Size:
Pack of 100
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15594789 100 Pack of 100
15643997 500 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15594789 Supplier BRAND™ Supplier No. 759125
  • ONLINE EXCLUSIVE PRICE
    51.70 €
    43.25 € / Pack of 100
    Save 8.45 €
    16% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BRAND™ UV-transparent cuvettes are the cost-effective alternative to glass or quartz cuvettes, perfect for numerous applications.

  • BRAND™ premium quality four-sided clear disposable cuvettes are tailored for superior fluorescence applications
  • Made of proprietary UV-plastic
  • Optimal for assays in the 230 to 800 nm range, offering virtually no autofluorescence for an excellent signal-to-noise ratio
  • Equipped with a 10 mm light path, these cuvettes are designed to fit most fluorimeters
TRUSTED_SUSTAINABILITY

Specifications

Sterility Non-sterile
Type Fluorimetry Cuvette
Description BRAND four sided clear cuvettes for fluorimetry. Proprietary UV-plastic. Package of 100
Color Transparent
Disposable Single-use
Capacity (Metric) 2.5 to 4.5 mL
Material UV-transparent Plastic
Length (Metric) 12.5mm
Includes Cuvette only
Quantity 100
Path Length 10 mm
Wavelength Range 230 to 800 nm
Height (Metric) 45 mm
For Use With (Equipment) Fluorimeter
Width (Metric) 12.5 mm
For Use With (Application) Fluorimetry
Lightpath 10 mm
Shape Rectangle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.