missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Four-Clear Sided Disposable Cuvettes for Fluorescence

Product Code. p-7146497 Shop All Brand Products
Click to view available options
Quantity:
100
500
Unit Size:
Pack of 100
Pack of 500
This item is not returnable. View return policy

Product Code. 15594789

Brand: BRAND™ 759125

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

BRAND™ UV-transparent cuvettes are the cost-effective alternative to glass or quartz cuvettes, perfect for numerous applications.

  • BRAND™ premium quality four-sided clear disposable cuvettes are tailored for superior fluorescence applications
  • Made of proprietary UV-plastic
  • Optimal for assays in the 230 to 800 nm range, offering virtually no autofluorescence for an excellent signal-to-noise ratio
  • Equipped with a 10 mm light path, these cuvettes are designed to fit most fluorimeters
TRUSTED_SUSTAINABILITY

Specifications

Sterility Non-sterile
Type Fluorimetry Cuvette
Description BRAND four sided clear cuvettes for fluorimetry. Proprietary UV-plastic. Package of 100
Color Transparent
Disposable Single-use
Capacity (Metric) 2.5 to 4.5 mL
Material UV-transparent Plastic
Length (Metric) 12.5mm
Includes Cuvette only
Quantity 100
Path Length 10 mm
Wavelength Range 230 to 800 nm
Height (Metric) 45 mm
For Use With (Equipment) flourometers
Width (Metric) 12.5 mm
For Use With (Application) Fluorimetry
Lightpath 10 mm
Shape Rectangle
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.