Learn More
BRAND™ Sealing paddle for sealing films
Product Code. 15708987 Shop All Brand Products
Change view
Click to view available options
Quantity:
2/Pk.
Unit Size:
Each
Product Code. Quantity Unit Size
15708987 2/Pk. Each
1 options
Product Code. 15708987 Supplier BRAND™ Supplier No. 701381
  • ONLINE EXCLUSIVE PRICE
    9.45 €
    6.55 € / Pack of 1
    Save 2.90 €
    30% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
for affixing plate sealing films
  • Assist in effective adhering of sealing films on microplates, PCR plates and deepwell plates
  • Package of 2
TRUSTED_SUSTAINABILITY
Specifications
Description Sealing paddle for affixing plate sealing films
For Use With (Equipment) Microplates, deep-well plates
No. per Pack 2/Pk.
Product Type Sealing Paddle
Quantity 2/Pk.