missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipette Tips, 0.1 - 20 μL, PP, Colorless
GREENER_CHOICE

Product Code. 15114937 Shop All Brand Products
Click to view available options
Format:
Rack
Refill
Tip Style:
Low Retention
Universal
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 15114937

Brand: BRAND™ 732322

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cleanroom quality for contamination-free analyses. Perfect fit for the Transferpette™ micropipette and many other pipettes. The tip system for all requirements.

Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 μl to 10 ml and filter tips for volumes from 0.1 μl to 1250 μl. They are tested for BRAND pipettes and many pipette from other manufacturers.

  • BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
  • Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
  • The nano-cap™ tip was specially developed for the nanoliter range and hence is ideal for molecular biological applications such as PCR. It is 37 mm long, and features a capillary rise that is even visible to the bare eye at 0.1 μl. The capillary part of the tip conveniently fills gel wells for gel electrophoresis systems from most manufacturers.
  • The racked tips are colorless and placed into a grey-colored tip tray.
  • The articles are available in the following quality levels: Certified Life Science Quality and BIO-CERT™ Liquid Handling Quality
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 20 μL
Sterility Sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 1.45 in.
Length (Metric) 3.7cm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Rack
Tip Style Low Retention
For Use With (Equipment) Transferpette S
Product Line Bio-Cert Nano-cap
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.