missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipette Tips, 0.1 - 20 μL, PP, Colorless

Product Code. 15124917 Shop All Brand Products
Change view
Click to view available options
Format:
Rack
Refill
Tip Style:
Low Retention
Universal
Unit Size:
Pack of 960
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Tip Style Unit Size
15124917 Rack Universal Pack of 960
15114907 Refill Universal Pack of 960
15114937 Rack Low Retention Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15124917 Supplier BRAND™ Supplier No. 732222
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleanroom quality for contamination-free analyses. Perfect fit for the Transferpette™ micropipette and many other pipettes. The tip system for all requirements.

Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 μl to 10 ml and filter tips for volumes from 0.1 μl to 1250 μl. They are tested for BRAND pipettes and many pipette from other manufacturers.

  • BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
  • Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
  • The nano-cap™ tip was specially developed for the nanoliter range and hence is ideal for molecular biological applications such as PCR. It is 37 mm long, and features a capillary rise that is even visible to the bare eye at 0.1 μl. The capillary part of the tip conveniently fills gel wells for gel electrophoresis systems from most manufacturers.
  • The racked tips are colorless and placed into a grey-colored tip tray.
  • The articles are available in the following quality levels: Certified Life Science Quality and BIO-CERT™ Liquid Handling Quality
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 20 μL
Sterility Sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 1.45 in.
Length (Metric) 3.7cm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Rack
Tip Style Universal
For Use With (Equipment) Transferpette S
Product Line Bio-Cert Nano-cap
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.