missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BIO-CERT™ Pipette Tips, 0.5 to 20 μL
GREENER_CHOICE

Product Code. 15134907 Shop All Brand Products
Change view
Click to view available options
Tip Style:
Low Retention, Narrow
Narrow
Format:
Box
Refill
Packaging:
TipBox, 10 boxes of 96 pieces
TipRack
TipRefill, 10 Racks x 96 Tips
TipStack, 10 Boxes x 96 Tips
Unit Size:
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tip Style Format Packaging unitSize
15134907 Narrow Refill TipRefill, 10 Racks x 96 Tips Pack of 960
15144917 Narrow Box TipRack Pack of 960
15124937 Low Retention, Narrow Box TipBox, 10 boxes of 96 pieces Pack of 960
15114927 Narrow Refill TipStack, 10 Boxes x 96 Tips Pack of 960
15124947 Low Retention, Narrow Refill TipStack, 10 Boxes x 96 Tips Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15134907 Supplier BRAND™ Supplier No. 732124
Wastewater Testing Savings
Use promo code 28023
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

DNA-, RNase-, ATP- and endotoxin-free

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Independently certified sterile and free from biological contaminants
  • Fits pipettes using ultra-micro/'crystal' tips
  • Slender design
  • Graduation at 2 and 10 μL for quick volume check
  • Available in either standard or ultra low retention types
  • Available in convenient, ecological rack-and-stack formats
  • Tips are colorless and placed into gray-colored tip tray

Ultra low retention tips

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface for low surface tension and low sample retention
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain proteins, nucleic acids, detergents and other difficult liquids that can cause sample loss
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging low retention properties
TRUSTED_SUSTAINABILITY

Specifications

Description BIO-CERT™ Liquid Handling Quality
Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Sterile
Color Colorless
Rack Color Gray
Autoclavable Autoclavable
Length (English) 1.81 in.
Length (Metric) 4.6 cm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) High translucency, easy ejection and low waste, quick volume check
Sterilization Method Beta Radiation
Format Refill
Tip Style Narrow
Packaging TipRefill, 10 Racks x 96 Tips
Product Type Pipet Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit Ultramicro
Graduated Yes
Product Line Bio-Cert
Material Polypropylene
Graduations 2, 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.