Learn More
BRAND™ PCR Cap Tool
Product Code. 15433105 Shop All Brand Products
Change view
Click to view available options
No. per Pack:
1 Ea.
Unit Size:
Each
Product Code. No. per Pack Unit Size
15433105 1 Ea. Each
1 options
Product Code. 15433105 Supplier BRAND™ Supplier No. 781419
  • ONLINE EXCLUSIVE PRICE
    48.65 €
    41.34 € / Pack of 1
    Save 7.31 €
    15% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with attaching and detaching caps on a PCR Plate.
  • Polyamide construction
  • Attaches and removes caps on PCR plates
TRUSTED_SUSTAINABILITY
Specifications
Compatibility All Plates Sealed with Strip Caps
For Use With (Application) Attaching and removing caps on a PCR plate
For Use With (Equipment) PCR plate caps
Material Polyamide
No. per Pack 1 Ea.
Product Type PCR Cap Tool