Learn More
BRAND™ nano-cap™ Pipet Tips
Product Code. 15144887 Shop All Brand Products
Click to view available options
:
Volume: 0.1 to 20 μL, Bulk
Product Code. 15144887
Brand: BRAND™ 732002
Only null left
Add to basket
Edge
Add to basket
Manufactured from high purity, DiHEMDA- and oleamide-free, virgin polypropylene. nano-cap™ Pipet Tips provide clean, hydrophobic surfaces, reduce sample retention, and increase accuracy and reproducibility.
  • Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene.
  • Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
  • Available either in bulk packages or in convenient rack-and-stack formats
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 20 μL
Sterility Non-sterile
Clean Claims DiHEMDA- and oleamide-free
Format Bulk
Product Line Nano-cap
Material Polypropylene
Graduations 20, 100μL