missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipette Tips, 0.5 to 20 μL

Product Code. 15114897 Shop All Brand Products
Change view
Click to view available options
Packaging:
Bulk
TipBox
TipBox, 5 boxes, 96 each
TipRack
TipStack, 2 x 5 refills, 96 each
Quantity:
10,000/Pk.
1000/Pk.
480/Pk.
960/Pk.
Filtered:
Non-filtered
Tip Style:
Low Retention, Narrow
Narrow
Unit Size:
Pack of 1000
Pack of 480
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity Filtered Tip Style Unit Size
15114897 Bulk 10,000/Pk. Non-filtered Narrow Pack of 1000
15164887 Bulk 1000/Pk. Non-filtered Narrow Pack of 1000
15104907 TipRack 960/Pk. Non-filtered Narrow Pack of 960
15184907 TipBox 480/Pk. Non-filtered Narrow Pack of 480
15174927 TipBox, 5 boxes, 96 each 480/Pk. - Low Retention, Narrow Pack of 480
15194917 TipStack, 2 x 5 refills, 96 each 960/Pk. - Narrow Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15114897 Supplier BRAND™ Supplier No. 732024
  • ONLINE EXCLUSIVE PRICE
    32.00 €
    29.10 € / Pack of 1000
    Save 2.90 €
    9% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from high-purity, oleamide and diHEMDA-free polypropylene. Pipet Tips, 0.5 to 20μL allow pipetting in microtubes and microplates without touching the walls. Gradations marked at 2μL and 10μL provide quick volume assessment.

  • Nonsterile , high-purity polypropylene is free from oleamide and diHEMDA.
  • Gradations marked at 2 μL and 10 μL provide quick volume assessment.
  • Slender design allows pipetting in microtubes and microplates without touching the walls.
  • Available in either standard or ultra low retention types.
  • Available either in bulk packages or in convenient rack-and-stack formats.

Ultra low retention tips

  • Produced through special, patented physicochemical process.
  • Homogeneous, defect-free surface thus produced has low surface tension.
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media.
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance.
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties.
TRUSTED_SUSTAINABILITY

Specifications

Description Life Science Quality
Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Length (English) 1.8 in.
Length (Metric) 46 mm
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) Reverse pipetting, plate washing, working with multichannel pipettes
Format Bulk
Tip Style Narrow
Packaging Bulk
Quantity 10,000/Pk.
Product Type Pipette Tips
Disposable Limited-use
For Use With (Equipment) Universal Fit Ultramicro
Graduated Yes
Material Polypropylene
Fit for Purpose PCR Ready
Graduations 2, 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.