Learn More
BrandTech™ BRAND™ Transferpette™ Single-Channel Pipetters: Tapered Pipet Shaft Cone
Product Code. 12181560
Change view
Click to view available options
Functional Range:
2 to 20μL, 0.5 to 10μL
Unit Size:
Each
Product Code. Functional Range Unit Size
12181560 2 to 20μL, 0.5 to 10μL Each
1 options
Product Code. 12181560 Supplier BrandTech™ Supplier No. 0101194
  • ONLINE EXCLUSIVE PRICE
    350.00 €
    338.00 € / Pack of 1
    Save 12.00 €
    3% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pipette shaft cone for Transferpette single channel pipettes, 10uL and 20uL using ultramicro tips.
  • Does not fit Transferpette S models
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 2 to 20μL digital ultra micro and 0.5 to 10μL Transferpette™ digital single channel pipette
Functional Range 2 to 20μL, 0.5 to 10μL
Includes Stainless Steel spring