missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Cuvette Caps

Product Code. 10086751 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Yellow
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10086751 Blue Pack of 100
10476221 Green Pack of 100
10139441 Orange Pack of 100
10159441 Yellow Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10086751 Supplier BRAND™ Supplier No. 759240
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides tight seal, allows storage of samples to -20°C.

  • Round cuvette caps
  • Fits ultra-micro cuvettes
  • Provide tight seal
  • Allows storage of samples to -20°
TRUSTED_SUSTAINABILITY

Specifications

Type Cuvette Cap
Color Blue
Disposable Single-use
Material Polyethylene
Quantity 100 Pack
For Use With (Equipment) Ultramicro UV Cuvette
Shape Round
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.