Learn More
BRAND™ QuikSip™ Bottle Top Aspirator Accessory, Manifold
Product Code. 10009790 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
QuikSip bottletop aspirator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10009790 QuikSip bottletop aspirator Each
1 options
Product Code. 10009790 Supplier BRAND™ Supplier No. 704526
  • ONLINE EXCLUSIVE PRICE
    14.00 €
    12.49 € / Pack of 1
    Save 1.51 €
    10% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Autoclavable and used withQuikSip™ Bottle Top Aspirator
  • 8-channel
  • Polypropylene
  • Autoclavable (121°C)
TRUSTED_SUSTAINABILITY
Specifications
Product Type 8 Channel Manifold
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Removal of fluids from multiwell plates
For Use With (Equipment) QuikSip bottletop aspirator