missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Screw Caps

Product Code. 10050571
Change view
Click to view available options
Capacity (Metric):
10,000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10050571 10,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050571 Supplier BRANDTECH™ VITLAB™ Supplier No. 100989
  • ONLINE EXCLUSIVE PRICE
    70.85 €
    60.30 € / Pack of 1
    Save 10.55 €
    14% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VITLAB™ polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Good chemical resistance for long-term storage of liquids.
  • Screw caps help protect against outside contamination and leakage.
  • Autoclavable at 121 C, according to DIN EN 285
TRUSTED_SUSTAINABILITY

Specifications

Capacity 338.1 oz., 10000mL
Capacity (Metric) 10,000 mL
Color Transparent
Diameter (English) 8.74 in.
Material Polypropylene
Quantity 6/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Threaded
Height (Metric) 394 mm
Height (English) 15.51 in.
Closure Material Polypropylene
Closure Size GL 63
Diameter (Metric) 222 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Capacity (English) 338.1 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.