missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Reagent Reservoirs

Product Code. 10327202 Shop All Brand Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
40 Bags of 5
Individually Wrapped
Packs of 10
Unit Size:
Pack of 10
Pack of 100
Pack of 200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Packaging unitSize
10327202 Sterile 40 Bags of 5 Pack of 200
10063970 Non-sterile Packs of 10 Pack of 10
10347392 Sterile Individually Wrapped Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10327202 Supplier BRAND™ Supplier No. 703409
Spotlight Offers
Use promo code 28036
View all promotions
In Stock
Add to basket
Edge
Add to basket

Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.

  • Heavy-gauge polypropylene
  • Suitable for all 8- and 12-channel pipetters
  • Autoclavable
  • Non-sterile version includes lid
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) For use with multichannel pipets. Without lid.
Packaging 40 Bags of 5
Quantity 200/Pk.
Autoclavable Autoclavable
Channels 1
Disposable Limited-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.