missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ Single Channel Electronic Pipets
GREENER_CHOICE

Product Code. 10150302 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 200 μL
100 to 1000 μL
2 to 20 μL
250 to 5000 μL
50 to 1000 μL
500 to 5000 μL
Color:
Blue
Gray
None
Red
Yellow
Includes:
EU plug
Europe AC Adapter
UK/Ireland AC Adapter
UK/Ireland plug
Without AC Adapter
Packungsgröße:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Volume (Metric) Color Includes unitSize
10150302 2 to 20 μL None Europe AC Adapter Each
10641603 0.5 to 10 μL Gray Without AC Adapter Each
10510882 10 to 200 μL Yellow Without AC Adapter Each
10120352 2 to 20 μL Gray Without AC Adapter Each
10102472 250 to 5000 μL Red Without AC Adapter Each
10511072 50 to 1000 μL Blue Without AC Adapter Each
12348593 500 to 5000 μL Red UK/Ireland AC Adapter Each
12338593 100 to 1000 μL Blue UK/Ireland AC Adapter Each
12318593 2 to 20 μL Gray UK/Ireland AC Adapter Each
12308593 0.5 to 10 μL - UK/Ireland plug Each
12328593 10 to 200 μL - UK/Ireland plug Each
10704733 10 to 200 μL - EU plug Each
10160302 50 to 1000 μL - EU plug Each
10485802 250 to 5000 μL - EU plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10150302 Lieferant BRAND™ Lieferanten-Nr. 705300

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

  • Functional, ergonomic housing design
  • Individually adjustable finger rest
  • Intuitive menu structure and comprehensively illustrated user manual
  • Significantly reduced tip attachment and ejection forces using universal tip
  • Corrosion-resistant piston and ejector
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
  • Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
  • Each pipette has a DE-M marking and is supplied with performance certificate
  • Supplied with battery and silicone oil
TRUSTED_SUSTAINABILITY

Spezifikation

Accuracy 0.2 μL (1.0%)
Battery Type Rechargeable NiMH
Certifications/Compliance DIN EN ISO 8655-2
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color None
Disposable Reusable
For Use With Pipette Tip (20 μL)
Material Polypropylene
Model DE-M
Number of Channels 1
Product Line Transferpette
Programmability Yes
Sterility Non-sterile
Type Singel Channel Microliter Pipette
Volume (Metric) 2 to 20 μL
Display LCD
Finger Style Ergonomic
Includes Europe AC Adapter
Quantity 1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.