missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ seripettor™ Bottletop Dispensers

Product Code. 10320861 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 mL
1 to 10 mL
2.5 to 25 mL
Increments:
0.04 mL
0.2 mL
0.5 mL
Viscosity Rating:
1000mm2/sec.
150mm2/sec.
75 mm2/sec.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Increments Viscosity Rating Unit Size
10320861 1 to 10 mL 0.2 mL 150mm2/sec. Each
10309744 0.2 to 2 mL 0.04 mL 1000mm2/sec. Each
10598882 2.5 to 25 mL 0.5 mL 75 mm2/sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10320861 Supplier BRAND™ Supplier No. 4720140
  • ONLINE EXCLUSIVE PRICE
    226.00 €
    172.30 € / Pack of 1
    Save 53.70 €
    23% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Bottle-top dispenser, for threaded bottles GL 45, includes discharge tube, filling tube, spare dispensing cartridge and PP adapters GL 32, GL 38 and S 40.

Suitable for use with bases, buffers, aqueous solutions of organic and inorganic substances, dilute non-oxidizing acids, and many polar solvents. Space-saving, low-rise design provides stability for easy one-handed operation and fit most common reagent bottles with the included adapters.

Suitable for use with bases, such as ammonia and sodium hydroxide; buffers; aqueous solutions of organic and inorganic substances; dilute non-oxidizing acids; and many polar solvents.

  • Fluid path consists of materials such as polyethylene, polypropylene and EPDM (ethylene-propylene-diene-rubber)
  • Full scale accuracy of ≤±1.2% and reproducibility of ≤0.2%
  • Fit most common reagent bottles with the included adapters
  • Space-saving, low-rise design provides stability for easy one-handed operation
  • With simple, clear volume adjustment for accuracy and a sturdy, supported discharge tube for safe operation
  • No fragile glass parts to break
  • Replaceable cylinder/piston cartridge has a wiping seal for renewability

This dispenser is not intended for use with scintillation fluid; non-polar solvents, such as xylene and most xylene substitutes; or strong, oxidizing or concentrated acids. For these liquids, choose the appropriate Dispensette™ bottletop dispenser.

Order Info

Unit disassembles in seconds for easy cleaning and maintenance. Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ≤±1.2%
Angle 90°
Product Type Seripettor Bottletop Dispenser
Autoclavable Autoclavable
Color Blue/Red/Clear
Density Up to 2.2 g/cm3
Channels 1
Material FEP, EPDM
Volume (Metric) 1 to 10 mL
Display Analog
For Use With (Application) Semi-automatic dispensing of alkaline, buffers, vitamin solutions, and nutrien media
Includes Discharge tube, telescoping filling tube, spare dispensing cartridge, 45/33mm and 45/38mm thread polypropylene adapters, operating manual
Increments 0.2 mL
Max. Pressure 500 mbar
Precision ≤0.2%
Temperature (English) 59°F to 104°F
Temperature (Metric) 15°C to 40°C
Vapor Pressure Up to 500 mbar
Viscosity Rating 150mm2/sec.
Warranty 1 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.