Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Product Code. 16254312
Change view
Click to view available options
For Use With:
MD 4C NT VARIO, PC3003/4 VARIO
ME 4C NT
ME 8C NT
MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
16254312 ME 4C NT Each
10404813 ME 8C NT Each
15960877 MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO Each
10355303 MD 4C NT VARIO, PC3003/4 VARIO Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16254312 Supplier VACUUBRAND™ Supplier No. 20696864
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
Service kits for VACUUBRAND NT series Chemistry design pumps.
  • Sets include appropriate diaphragms, valves and sealing rings (if applicable)
  • Wrench not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With ME 4C NT
Type Service Kit