Learn More
VACUUBRAND™ Supplementary Kit Connection for BVC Fluid Aspiration Systems
Product Code. 12695640
Change view
Click to view available options
For Use With:
BVC fluid aspiration system
Unit Size:
1 set
Product Code. For Use With unitSize
12695640 BVC fluid aspiration system 1 set
1 options
Product Code. 12695640
Brand: VACUUBRAND™ 20699943
Only null left
Add to basket
Add to basket
Replacement parts and accessories for VACUUBRAND BVC fluid aspiration systems
The BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work. They provide the flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in the bottle.
  • To add second VHCPRO to BVC bottle aspiration cap
TRUSTED_SUSTAINABILITY
Specifications
For Use With BVC fluid aspiration system
Type Supplementary Kit Connection