missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Transferpette™ S Multichannel Pipet

Product Code. 10446513 Shop All Brand Products
Change view
Click to view available options
Accuracy:
±0.6%
±0.8%
±1.6%
Number of Channels:
8
12
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Accuracy Number of Channels Volume (Metric) unitSize
10446513 ±0.8% 12 10 to 100 μL Each
10161393 ±0.8% 8 5 to 50 μL Each
10610794 ±1.6% 12 0.5 to 10 μL Each
10571905 ±0.8% 12 5 to 50 μL Each
10796164 ±0.8% 12 20 to 200 μL Each
10406333 ±0.8% 8 20 to 200 μL Each
10796354 ±0.6% 12 30 to 300 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10446513 Supplier BRAND™ Supplier No. 703728

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual

Protects against accidental volume changes using innovative one-handed volume adjustment with brake. Supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty.

Protects against accidental volume changes using innovative one-handed volume adjustment with brake. BrandTech™ Transferpette™ S Multichannel Pipet is supplied with performance certificate, operating manual, shelf mount, two tip boxes, reagent reservoir, pack of replacement v-rings, and one year warranty. Patented individually removable/replaceable tip cones provide easy in-lab cleaning.

Device has one-handed volume adjustment with brake for secure setting.

  • With central button control layout
  • Short, 12.5mm piston stroke-length
  • V-rings for low-force tip mounting and ejection
  • Four digit volume display
  • Lightweight
  • Individually removable/replaceable patented tip cones
  • Autoclavable at 121°C (20 minutes)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.8%
Autoclavable Autoclavable
Number of Channels 12
Product Line Transferpette S
Type Pipette
Volume (Metric) 10 to 100 μL
Display Digital
Includes Shelf/rack mount, box of pipette tips with two refills, set of replacement V-rings, certificate of performance and comprehensive operating manual
Stroke (Metric) Length 12.5mm
Warranty 1 Year
Dimensions (L x W x H) 241.3 x 142.8 x 123.8mm
Length (English) 9.5 in.
Length (Metric) 241.3 mm
Width (English) 5.62 in.
Width (Metric) 142.8 mm
Height (English) 4.87 in.
Height (Metric) 123.8 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.