Learn More
BRAND™ Transferpette™ S Adjustable Single Channel Pipet Starter Sets
Product Code. 10179002 Shop All Brand Products
Click to view available options
Volume (Metric):
0.1 to 1, 0.5 to 10, 10 to 100 μL
0.5 to 10, 10 to 100, 100 to 1000 μL
100 to 1000, 500 to 5000, 1000 to 10,000 μL
2 to 20, 20 to 200, 100 to 1000 μL
Includes:
D-1, D-10, D-100 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-10, D-100, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
D1000, D5000, D10000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes
Type:
Macro Set
Micro Set
Pipette Set, Standard
Starter Set, Adjustable
Unit Size:
Each
Product Code. 10179002
Brand: BRAND™ 704791
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Set contents:
  • Three adjustable volume pipettes
  • Three shelf/rack mounts
  • Three filled polypropylene Tip Boxes
Simple one-handed operation units exhibits high degree accuracy, precision, maximum versatility and reliable performance with Easy Calibration™ technology
  • Available as Standard Set, Micro Set, Midsize Set and Macro Set
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With Laboratory
Number of Channels 1
Product Line Transferpette
Type Starter Set, Adjustable
Volume (Metric) 2 to 20, 20 to 200, 100 to 1000 μL
For Use With (Application) Demanding laboratory applications
Includes D-20, D-200, D-1000 adjustable volume pipette, three shelf/rack mounts, three filled Polypropylene tip boxes