Learn More
Branson Ultrasonics™ Sonifier™ Replacement Flat Tip
Product Code. 10337832
Change view
Click to view available options
For Use With (Equipment):
Sonifier™ cell disruptor
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
10337832 Sonifier™ cell disruptor Each
1 options
Product Code. 10337832 Supplier Branson Ultrasonics™ Supplier No. 101148013
  • ONLINE EXCLUSIVE PRICE
    106.05 €
    84.00 € / Pack of 1
    Save 22.05 €
    20% Off
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tip for 1/2 in. probe.
Tip to use as a 1/2 in. probe, without tip probe used to attach tapered microtips.
Specifications
Product Type Flat Microtip
For Use With (Equipment) Sonifier™ cell disruptor
Tip Style Flat