missing translation for 'onlineSavingsMsg'
Learn More

Branson Ultrasonics™ Soundproof Enclosure

Product Code. 10468481
Change view
Click to view available options
Length (Metric):
31.8 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10468481 31.8 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10468481 Supplier Branson Ultrasonics™ Supplier No. 101063275
  • ONLINE EXCLUSIVE PRICE
    1547.87 €
    1220.00 € / Pack of 1
    Save 327.87 €
    21% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Requires

Adapter 15-335-300 (only with 150 watt Sonifier)

For use with Sonifier™ Cell Disruptors

Minimizes mechanical noise produced by ultrasonic processing.

  • Lowers noise levels by 20-25dBA
  • Sturdy cabinet is lined with waterproof, sound-abating material that is impervious to most solutions and reagents and cleans easily
  • See-through door allows process viewing
  • Opens at top and bottom for easy access with a cup horn or standard probes

Order Info

Interior dimensions: 10.5L x12.75W x 20.75 in. H (26.7 x 32.4 x 52.7cm). Overall dimensions: 12L x 14.5W x 23 in. H (30.5 x 36.8 x 58.4cm). Net Weight: 19.5 lb. (8.8kg).

Specifications

Dimensions (L x W x H) 12.5 x 18.5 x 28 in. (31.8 x 47 x 71.1 cm)
Height (English) 28 in.
Height (Metric) 71.1 cm
Length (English) 12.5 in.
Length (Metric) 31.8 cm
Width (English) 18.5 in.
Width (Metric) 47 cm
For Use With (Application) Ideal for minimizes mechanical noise levels by 20 to 25dBA produced by ultrasonic processing
For Use With (Equipment) Sonifier™ cell disruptor
Product Type Soundproof Enclosure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.