Learn More
Brass foil, alloy 260, 0.25mm (0.01in) thick, 15x45cm (5.9x18in), Thermo Scientific Chemicals
71.10 € - 252.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426015
|
Thermo Scientific Alfa Aesar
013503.KJ |
5 Pc. |
252.00 €
Pack of 5 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15416015
|
Thermo Scientific Alfa Aesar
013503.KG |
1 Pc. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Brass foil, alloy has a wide range of applications, such as appliance parts, highly formable parts, ammunition components, decorative parts of furniture and cabinets, name plates, engraving plates, overlays, legend plates, lamp fixtures, fasteners, locks, hinges, flashlight shells, connectors, fittings, circuit board relays, electronic components, switches, nuts, bolts, valve stems and valve components, radiator cores and tanks, brackets, frames, fixtures, ornamental works, and used extensively in the musical instrument, electrical equipment, plumbing, central heating, seawater lines and desalination fields.