missing translation for 'onlineSavingsMsg'
Learn More

Brass spheres, alloy 260, 6.35mm (0.25in) dia

Product Code. 15496785
Change view
Click to view available options
Quantity:
100 Pc.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496785 100 Pc. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496785 Supplier Thermo Scientific Alfa Aesar Supplier No. 045162.KN
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-50-8 | Cu | 63.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-50-8
Appearance Copper colored spheres
Quantity 100 Pc.
Odor Odorless
Form Sphere
Molecular Formula Cu
MDL Number MFCD00010965
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Chemical Name or Material Brass
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.