missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD4 degrader AT1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
BRD4 degrader AT1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 as a highly selective Brd4 degrader, with a Kd of 44 nM for Brd4BD2 in cells.
Specifications
Specifications
| Chemical Name or Material | BRD4 degrader AT1 |
| CAS | 2098836-45-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C48H58ClN9O5S3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (102.81 mM; Need ultrasonic) |
| SMILES | CC1=C(C)SC2=C1C(C3=CC=C(Cl)C=C3)=N[C@@H](CC(NCCCCCCSC(C)(C)[C@H](NC(C)=O)C(N4[C@@H](C[C@@H](O)C4)C(NCC5=CC=C(C6=C(C)N=CS6)C=C5)=O)=O)=O)C7=NN=C(C)N27 |
| Molecular Weight (g/mol) | 972.68 |
| Formula Weight | 972.68 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction