missing translation for 'onlineSavingsMsg'
Learn More

BRD4 degrader AT1, MedChemExpress

Product Code. p-200098448
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447177

Brand: Medchem Express HY111433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BRD4 degrader AT1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 as a highly selective Brd4 degrader, with a Kd of 44 nM for Brd4BD2 in cells.

BRD4 degrader AT1 is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 as a highly selective Brd4 degrader, with a Kd of 44 nM for Brd4BD2 in cells.

Specifications

Chemical Name or Material BRD4 degrader AT1
CAS 2098836-45-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C48H58ClN9O5S3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (102.81 mM; Need ultrasonic)
SMILES CC1=C(C)SC2=C1C(C3=CC=C(Cl)C=C3)=N[C@@H](CC(NCCCCCCSC(C)(C)[C@H](NC(C)=O)C(N4[C@@H](C[C@@H](O)C4)C(NCC5=CC=C(C6=C(C)N=CS6)C=C5)=O)=O)=O)C7=NN=C(C)N27
Molecular Weight (g/mol) 972.68
Formula Weight 972.68
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.