missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bremelanotide Acetate Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Bremelanotide Acetate Hydrate |
| Purity Grade Notes | HPLC |
| Molecular Formula | C52H74N14O13 |
| Quantity | 100 mg |
| Synonym | N-Acetyl-L-norleucyl-L-α-aspartyl-L-histidyl-D-phenylalanyl-L-arginyl-L-tryptophyl-L-Lysine (2→7)-Lactam Acetate Hydrate 3: PN: WO2006060873 SEQID: 3 Claimed Protein Acetate Hydrate, Bremelanotide Acetic Acid, PT 141 Acetate Hydrate |
| InChI Formula | InChI=1S/C50H68N14O10.C2H4O2.H2O/c1-3-4-16-35(58-29(2)65)43(67)64-41-25-42(66)54-20-11-10-18-37(49(73)74)60-46(70)39(23-31-26-56-34-17-9-8-15-33(31)34)62-44(68)36(19-12-21-55-50(51)52)59-45(69)38(22-30-13-6-5-7-14-30)61-47(71)40(63-48(41)72)24-32-27-53-28-57-32;1-2(3)4;/h5-9,13-15,17,26-28,35-41,56H,3-4,10-12,16,18-25H2,1-2H3,(H,53,57)(H,54,66)(H,58,65)(H,59,69)(H,60,70)(H,61,71)(H,62,68)(H,63,72)(H,64,67)(H,73,74)(H4,51,52,55);1H3,(H,3,4);1H2/t35-,36+,37-,38+,39+,40+,41-;;/m0../s1 |
| SMILES | O=C([C@@H](CC1=CC=CC=C1)NC([C@@H](CC2=CN=CN2)NC([C@@H](NC([C@H](C[C]CC)NC(C)=O)=O)CC(NCCCC[C@@H](C(O)=O)NC3=O)=O)=O)=O)N[C@H](CCCNC(N)=N)C(N[C@@H]3CC4=CNC5=C4C=CC=C5)=O.OC(C)=O.O |
| IUPAC Name | (3R,6R,9R,12R,15S,23S)-15-[[(2S)-2-acetamidohexanoyl]amino]-9-benzyl-6-[3-(diaminomethylideneamino)propyl]-12-(1H-imidazol-5-ylmethyl)-3-(1H-indol-3-ylmethyl)-2,5,8,11,14,17-hexaoxo-1,4,7,10,13,18-hexazacyclotricosane-23-carboxylic acid;acetic acid;hydrate |
| Molecular Weight (g/mol) | 1103.23 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?