missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 30

Product Code. 10591154 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10591154 2.5 kg 2.5kg
10134510 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591154 Supplier Thermo Scientific Acros Supplier No. 216720025
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene(4)lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color Colorless to Yellow
Acid Value 2mg KOH/g max.
Physical Form Liquid
Melting Point 14.0°C
Density 0.9500g/mL
Quantity 2.5 kg
Flash Point >149°C
Hydroxyl Value 145 to 165mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Specific Gravity 0.95
Viscosity 30 mPa.s (25°C)
Linear Formula C12H25(OCH2CH2)4OH
Water 1% max.
Fieser 01,892
Merck Index 15, 7675
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in lower alcohols, propylene glycol, insoluble in mineral oil, cottonseed oil
Formula Weight 362.54
Chemical Name or Material Brij™ 30
Show More Show Less
compliance-icons
Product Identifier
  • Brij|TS 30
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.