missing translation for 'onlineSavingsMsg'
Learn More

Brij™ L23-SO-(AP), 30 w/v % sol. in water, previous name Brij™ 35

Product Code. 10318653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10318653 2.5 L 2.5 L
10265731 100 mL 100mL
10798172 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318653 Supplier Thermo Scientific Acros Supplier No. 329580025
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-92-0 | C58H118O24

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brij™ 35
Color Colorless
CAS 9002-92-0, 7732-18-5
Physical Form Jelly Substance at 25°C
Density 1.0300g/mL
Quantity 2.5 L
Packaging Glass bottle
Specific Gravity 1.03
Molecular Formula C58H118O24
Linear Formula C12H25(OCH2CH2)23OH
MDL Number MFCD00080891
Fieser 06,70
Merck Index 15, 7675
Solubility Information Solubility in water: soluble.
Formula Weight 1199.56
Percent Purity 29 to 31%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.