missing translation for 'onlineSavingsMsg'
Learn More

Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35

Product Code. 11956941 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
HDPE plastic bottle
Plastic drum
Unit Size:
1kg
250g
500g
5kg
This item is not returnable. View return policy

Product Code. 11956941

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene(23)lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color White
Acid Value 5mg KOH/g max.
Physical Form Waxy Pellets
Boiling Point >100.0°C
Melting Point 33.0°C to 40.0°C
Quantity 5 kg
Flash Point >149°C
Hydroxyl Value 40 to 60mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic drum
Linear Formula C12H25(OCH2CH2)23OH
Water 1% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils
Formula Weight 1199.56
Chemical Name or Material Brij™ 35
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.