missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brij™-35 Surfact-Amps™ Detergent Solution

Product Code. 13414269 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
6 x 10 mL
Unit Size:
500mL
50mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13414269 50 mL 50mL
11841455 6 x 10 mL Pack of 6
13424269 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 13414269 Supplier Thermo Scientific™ Supplier No. 85117
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.

Thermo Scientific™ Brij-35 Detergent Solutions are stabilized 10% and 30% solutions of Brij-35 detergent for use in various protein methods.

Additional Information and Online Ordering

Brij-35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or as a surfactant in various HPLC applications. The detergent is offered in two forms: a Thermo Scientific Surfact-Amps™ Detergent Solution (high-purity, 10% solution) and a standard grade 30% solution.

Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neet detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Highlights:

  • Brij-35 – a nonionic detergent for use in various protein methods

  • Accurate – 10% and 30% detergent solutions in ultrapure water

  • Easy-to-use – solutions are simple to dispense and dilute for use

Detergent Properties:

  • Molecular Weight: 1225g
  • Detergent Class: Nonionic
  • Aggregation Number: 40
  • Micelle Molecular Weight: 49,000g
  • Critical Micelle Concentration (CMC): 0.09mM (0.011%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Surfact-Amps Detergent Solution (Part No. 28316):

  • Accurate – precise 10% detergent solution in ultrapure water

  • Easy-to-use – solution is simple to dispense and dilute for use

  • Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls

  • Stable – packed in glass ampules under inert nitrogen gas

TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store at Room Temperature
Form Liquid
Product Line Brij, Surfact-Amps
Reagent Type Detergent Solution
Quantity 50 mL
compliance-icons
Product Identifier
  • Brij-35 Surfact-Amps Detergent Solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Ethoxylated lauryl alcohol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.