missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 35, Fisher BioReagents

Product Code. 10366553
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10366553 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366553 Supplier Fisher BioReagents Supplier No. BP345500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This nonionic detergent is used for solubilizing proteins and isolating membrane complexes.
TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9002-92-0
Physical Form Solid
Melting Point 40°C
pH 5.5 to 7.0
Quantity 500 g
Hydroxyl Value 40 to 60
Identification Pass Test
Packaging Glass Bottle
Molecular Formula C14H30O2
Linear Formula C12H25 (OCH2CH2)n
Water 3% max.
MDL Number MFCD00043063
Merck Index 15, 7675
Synonym Polyoxyethylene (23) Lauryl Ether
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCOCCO
IUPAC Name 2-(dodecyloxy)ethan-1-ol
Molecular Weight (g/mol) 230.39
Formula Weight 1199.56
Chemical Name or Material Brij™ 35
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.