missing translation for 'onlineSavingsMsg'
Learn More

BRIJ™ 35, MP Biomedicals™

Product Code. 11406601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406601 250 g 250g
11496591 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406601 Supplier MP Biomedicals Supplier No. 0219484083
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene 23 Lauryl Ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Physical Form Powder
Quantity 250 g
Chemical Name or Material BRIJ 35
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BRIJ 35, molecular biology reagent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.