missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 98

Product Code. 10117553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10117553 100 g Glass bottle 100g
10641832 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117553 Supplier Thermo Scientific Acros Supplier No. 347181000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-98-2 | C58H116O21 | 1149.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym Polyoxyethylene(20) oleyl ether
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color White to Cream
Acid Value 1mg KOH/g max.
Physical Form Soft Waxy Solid
Boiling Point >100.0°C
Melting Point 30.0°C
Density 1.0700g/mL
Quantity 100 g
Flash Point >150°C
Hydroxyl Value 50 to 65mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.07
Linear Formula C18H35(OCH2CH2)nOH
Water 3% max.
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, toluene, propylene glycol, insoluble in cottonseed oil, mineral oil
Formula Weight 1149.56
Chemical Name or Material Brij™ 98
Show More Show Less
compliance-icons
Product Identifier
  • Brij|TS 98
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.