missing translation for 'onlineSavingsMsg'
Learn More

Brij L23, TRC

Product Code. 30428645
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428645 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428645 Supplier Toronto Research Chemicals Supplier No. TRCP68827510G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C58H118O24
Molecular Weight (g/mol) 1199.54
Synonym Brij™ 35 Tricosaethylene Glycol Mono-n-dodecyl Ether
SMILES CCCCCCCCCCCC[(OCH2CH2)nOH]

Specifications

Quantity 10 g
Formula Weight 1198.8
Chemical Name or Material Polyoxyl Lauryl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.