missing translation for 'onlineSavingsMsg'
Learn More

Brij∣r L23, Electrophoresis Reagent

Product Code. 15467329
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467329 250 g 250g
15457329 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467329 Supplier Thermo Scientific Alfa Aesar Supplier No. J66096.30
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-92-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brij™ L23
Color White
Name Note Electrophoresis Reagent
CAS 9002-92-0
Physical Form Waxy Solid
Melting Point ∼35°C to 40°C
Quantity 250 g
MDL Number MFCD00080891
Synonym Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether
Solubility Information Soluble in water
Formula Weight 1199.57
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.