missing translation for 'onlineSavingsMsg'
Learn More

Brij∣r L23

Product Code. 11442478
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442478 5000 g 5000g
11432478 250 g 250g
11422478 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442478 Supplier Thermo Scientific Alfa Aesar Supplier No. A15809.0I
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol

Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).

Solubility
Soluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.

Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Melting Point ∼35°C to 40°C
Quantity 5000 g
Linear Formula CH3(CH2)10CH2(OCH2CH2)23OH
Solubility Information Soluble in water (40mg/ml) at 20°C methanol,chloroform and ethanol.
Formula Weight 1199.57
Chemical Name or Material Brij™ L23
compliance-icons
Product Identifier
  • Brij L23
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.