missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Black, MP Biomedicals

Product Code. 11461046 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461046 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11461046 Supplier MP Biomedicals Supplier No. 0520232710
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Enables simultaneous isolation of high-quality DNA and RNA from soil samples with effective removal of PCR inhibitors.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2519-30-4
Molecular Formula C28H17N5Na4O14S4
InChI Key AGKKBBSOKGLVTM-YENKNCRGSA-J
SMILES CC(=O)NC1=C2C(=C(C=C1)S(=O)(=O)[O-])C=C(C(=NNC3=C4C=C(C=CC4=C(C=C3)N=NC5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-])C2=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
IUPAC Name tetrasodium;(6E)-4-acetamido-5-oxo-6-[[7-sulfonato-4-[(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]hydrazinylidene]naphthalene-1,7-disulfonate
Molecular Weight (g/mol) 867.664
PubChem CID 6321379
Formula Weight 871.74
Color Black
Quantity 10 g
Chemical Name or Material Brilliant Black
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.