missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Brilliant Blue FCF

Product Code. 16442429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442429 Supplier MedChemExpress Supplier No. HYD0915

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Brilliant Blue FCF has the appearance of a reddish-blue powder.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Synonym Acid Blue 9 ∣ FD & C Blue No. 1 ∣ E133
CAS 3844-45-9
Molecular Weight (g/mol) 792.85 g/mol
Solubility H2O : 33.33 mg/mL (42.04 mM; ultrasonic and adjust pH to 11 with NaOH)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.