missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Green

Product Code. 11499010
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499010 25 g 25g
11409020 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499010 Supplier Thermo Scientific Chemicals Supplier No. A12801.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 633-03-4 | C27H34N2O4S | 482.639 g/mol

Brilliant Green a stain used in bacteriological media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Green a stain used in bacteriological media.

Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).

Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
MDL Number MFCD00011880
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Basic Green 1; C.I. 42040
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]

Specifications

Melting Point 210°C (decomposition)
Beilstein 3901207
Merck Index 14,1374
Solubility Information Soluble in water (100mg/ml at 25°C),ethanol (50mg/ml),methanol,and 2-methoxyethanol (60mg/ml).
Formula Weight 482.65
Quantity 25 g
Chemical Name or Material Brilliant Green
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Brilliant Green
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : C.I. Basic Green 1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.