missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant Green, Certified, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| CAS | 633-03-4 |
| Absorption Ratio | 0.94 to 1.14 |
| Dye Content | 85% min. |
| Molecular Formula | C27H34N2O4S |
| MDL Number | MFCD00011880 |
| Beilstein | 13, 746 |
| Merck Index | 15, 1380 |
| Synonym | Basic Green 1, C.I. 42040, Ethyl Green |
| InChI Key | NNBFNNNWANBMTI-UHFFFAOYSA-M |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
Product Identifier
- Brilliant Green, high purity biological stain
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction