missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant Green, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Description
Brilliant green is used in dilute solution as a topical antiseptic. It is effective against gram-positive microorganisms. It has also been used to dye silk and wool.
Specifications
Specifications
| CAS | 633-03-4 |
| Molecular Formula | C27H34N2O4S |
| Synonym | Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO |
| Solubility Information | Soluble in water (1mg/mL), ethanol, methanol. |
| InChI Key | NNBFNNNWANBMTI-UHFFFAOYSA-M |
| SMILES | CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-] |
| IUPAC Name | [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate |
| Molecular Weight (g/mol) | 482.639 |
| PubChem CID | 12449 |
| Formula Weight | 482.65 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction