missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant green, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 633-03-4 |
| Molecular Formula | C27H34N2O4S |
| Synonym | Basic green l, C.I. 42040 |
| InChI Key | NNBFNNNWANBMTI-UHFFFAOYSA-M |
| SMILES | CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-] |
| IUPAC Name | [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate |
| Molecular Weight (g/mol) | 482.639 |
| PubChem CID | 12449 |
| Formula Weight | 482.65 |
| Color | Green |
| Show More |
Safety and Handling
Product Identifier
- Brilliant Green
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Brilliant Green
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction