missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Brilliant Stain Buffer

Product Code. 17470683
Change view
Click to view available options
Quantity:
100 Tests
1000 Tests
Unit Size:
100 tests
1000 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17470683 1000 Tests 1000 tests
17460683 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17470683 Supplier Invitrogen™ Supplier No. 00440975
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension. Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample.

  • For use as a supplement to Flow Cytometry Staining Buffer in immunofluorescent staining protocols of cells in suspension
  • Brilliant Stain Buffer is only necessary when using more than one polymer dye-conjugated antibody in the same sample
  • Prevents non-specific polymer interactions that can result in data appearing under-compensated
  • Buffer is provided in a 50 μL/test format
  • Compatible with traditional fluorochromes, Brilliant Violet dyes, Brilliant Ultra Violet dyes, Super Bright dyes, and standard flow cytometry protocols
  • Buffer is not compatible with UltraComp eBeads and UltraComp eBeads Plus compensation beads
  • If using these beads as a compensation tool, Flow Cytometry Staining Buffer should be used exclusively for all dilutions and washes during staining of the microspheres
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Staining Buffer
Quantity 1000 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.