missing translation for 'onlineSavingsMsg'
Learn More

Brimonidine L-Tartrate, TRC

Product Code. 30347944
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347944 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347944 Supplier Toronto Research Chemicals Supplier No. TRCB677530100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brimonidine tartrate
CAS 70359-46-5
Molecular Formula C11 H10 Br N5 . C4 H6 O6
Quantity 100 mg
Synonym Brimonidine Tartrate, 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine (2R,3R)-2,3-dihydroxybutanedioate (1:1), 5-Bromo-N-(4,5-dihydro-1H-imidazol-2-yl)-6-quinoxalinamine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), AGN 190342LF, Alphagan, Alphagan P, Brimonidine Purite, Brimonidine tartrate, Brominide tartrate
InChI Formula InChI=1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
SMILES O[C@H]([C@@H](O)C(=O)O)C(=O)O.Brc1c(ccc2nccnc12)N=C3NCCN3
IUPAC Name N-(5-bromoquinoxalin-6-yl)imidazolidin-2-imine;(2R,3R)-2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 442.22
Formula Weight 441.0284
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.