missing translation for 'onlineSavingsMsg'
Learn More

Brinzolamide Hydrochloride, TRC

Product Code. 30004515
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004515 10 mg 10mg
30133202 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004515 Supplier Toronto Research Chemicals Supplier No. TRCB67760010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150937-43-2
Formula Weight 419.041
InChI Formula InChI=1S/C12H21N3O5S3.ClH/c1-3-14-10-8-15(5-4-6-20-2)23(18,19)12-9(10)7-11(21-12)22(13,16)17;/h7,10,14H,3-6,8H2,1-2H3,(H2,13,16,17);1H/t10-;/m0./s1
IUPAC Name (4R)-4-(ethylamino)-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide;hydrochloride
Molecular Formula C12 H21 N3 O5 S3 . Cl H
Molecular Weight (g/mol) 419.97
SMILES Cl.CCN[C@H]1CN(CCCOC)S(=O)(=O)c2sc(cc12)S(=O)(=O)N
Synonym Brinzolamide Hydrochloride
Chemical Name or Material Brinzolamide Hydrochloride

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.