Learn More
Fisherbrand™ Ion Selective Electrode
Product Code. 11715648 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Membrane:
Combination
Unit Size:
Each
Product Code. Membrane Unit Size
11715648 Combination Each
1 options
Product Code. 11715648
  • ONLINE EXCLUSIVE PRICE
    479.00 €
    302.17 € / Pack of 1
    Save 176.83 €
    36% Off
View all promotions
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Combined ion-sensitive bromide electrode with solid membrane interchangeable flat surface is ideally delivered with membrane, electrolytic solution and gasket.
  • Combined ion-sensitive bromide electrode with solid membrane interchangeable flat surface
  • The ion to be detected passes through the membrane generating a potential difference proportional to the ionic concentration
  • Analysis time typically less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
Specifications
Description Bromide Ion Selective Electrode
pH Range 2 to 14
Membrane Combination