Learn More
Proteintech Broad Range Prestained Protein Marker
The PL00002 Broad Range Protein Marker is a ready-to-use three-color protein standard with 13 prestained proteins covering a wide range of molecular weights from 3 to 245 kDa in Tris-Glycine buffer.
Brand:  Proteintech PL00002-500UL
Product Code. 17318262
  • 139.00 € / 500µL
Description
  • The PL00002 Broad Range Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes (nitrocellulose, PVDF, or nylon) and for estimating the size of proteins.
  • Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).