missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromadiolone-d5 (phenyl-d5) (mixture of diastereomers), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Bromadiolone-d5 (Mixture of Diasteromers) |
| Molecular Formula | C30 D5 H18 Br O4 |
| Quantity | 0.01 g |
| Synonym | Bromadiolone-d5 (phenyl-d5) (mixture of diastereomers), Bromadiolone-D5 (phenyl-D5), 3-[α-[p-(p-Bromophenyl)-β-hydroxyphenethyl]benzyl-2,3,4,5,6-d5]-4-hydroxycoumarin, 3-[3-(4'-Bromo[1,1'-biphenyl]-4-yl)-3-hydroxy-1-(phenyl-d5)propyl]-4-hydroxy-2 H-1-benzopyran-2-one |
| InChI Formula | InChI=1 S/C30H23BrO4/c31-23-16-14-20(15-17-23)19-10-12-22(13-11-19)26(32)18-25(21-6-2-1-3-7-21)28-29(33)24-8-4-5-9-27(24)35-30(28)34/h1-17,25-26,32-33 H,18H2/i1D,2 D,3 D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(CC(O)c2ccc(cc2)c3ccc(Br)cc3)C4=C(O)c5ccccc5OC4=O)c([2 H])c1[2 H] |
| IUPAC Name | 3-[3-[4-(4-bromophenyl)phenyl]-3-hydroxy-1-(2,3,4,5,6-pentadeuteriophenyl)propyl]-4-hydroxychromen-2-one |
| Molecular Weight (g/mol) | 532.436 |
| Formula Weight | 531.1094 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction